Learn More
Hampton Research HR2-928-D3/Grid Screen PEG/LiCl #D3, 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Lithium chloride, 30% w/v Polyethylene glycol 6,000
SDP
Catalog No. 501940
Encompass
HR2-928-D3/Grid Screen PEG/LiCl #D3, 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Lithium chloride, 30% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-194-0 Supplier Hampton Research Supplier No. HR2928D3
HR2-928-D3/Grid Screen PEG/LiCl #D3, 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Lithium chloride, 30% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.