Learn More
Hampton Research HR2-932-A2/Grid Screen Sodium Chloride #A2/Grid Screen Salt HT (G02), 185 ml - 0.1 M Citric acid pH 5.0, 1.0 M Sodium chloride
SDP
Catalog No. 501984
Encompass
HR2-932-A2/Grid Screen Sodium Chloride #A2/Grid Screen Salt HT (G02), 185 ml - 0.1 M Citric acid pH 5.0, 1.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-198-4 Supplier Hampton Research Supplier No. HR2932A2
HR2-932-A2/Grid Screen Sodium Chloride #A2/Grid Screen Salt HT (G02), 185 ml - 0.1 M Citric acid pH 5.0, 1.0 M Sodium chloride. Please note items from this supplier are non-returnable.