Learn More
Hampton Research HR2-932-A3/Grid Screen Sodium Chloride #A3/Grid Screen Salt HT (G03), 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Sodium chloride
SDP
Catalog No. 501985
Encompass
HR2-932-A3/Grid Screen Sodium Chloride #A3/Grid Screen Salt HT (G03), 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-198-5 Supplier Hampton Research Supplier No. HR2932A3
HR2-932-A3/Grid Screen Sodium Chloride #A3/Grid Screen Salt HT (G03), 185 ml - 0.1 M MES monohydrate pH 6.0, 1.0 M Sodium chloride. Please note items from this supplier are non-returnable.