Learn More
Hampton Research HR2-932-B4/Grid Screen Sodium Chloride #B4/Grid Screen Salt HT (G10), 185 ml - 0.1 M HEPES pH 7.0, 2.0 M Sodium chloride
SDP
Catalog No. 501992
Encompass
HR2-932-B4/Grid Screen Sodium Chloride #B4/Grid Screen Salt HT (G10), 185 ml - 0.1 M HEPES pH 7.0, 2.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-199-2 Supplier Hampton Research Supplier No. HR2932B4
HR2-932-B4/Grid Screen Sodium Chloride #B4/Grid Screen Salt HT (G10), 185 ml - 0.1 M HEPES pH 7.0, 2.0 M Sodium chloride. Please note items from this supplier are non-returnable.