Learn More
Hampton Research HR2-932-D3/Grid Screen Sodium Chloride #D3/Grid Screen Salt HT (H09), 185 ml - 0.1 M MES monohydrate pH 6.0, 4.0 M Sodium chloride
SDP
Catalog No. 502003
Encompass
HR2-932-D3/Grid Screen Sodium Chloride #D3/Grid Screen Salt HT (H09), 185 ml - 0.1 M MES monohydrate pH 6.0, 4.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-200-3 Supplier Hampton Research Supplier No. HR2932D3
HR2-932-D3/Grid Screen Sodium Chloride #D3/Grid Screen Salt HT (H09), 185 ml - 0.1 M MES monohydrate pH 6.0, 4.0 M Sodium chloride. Please note items from this supplier are non-returnable.