Learn More
Hampton Research HR2-932-B6/Grid Screen Sodium Chloride #B6/Grid Screen Salt HT (G12), 185 ml - 0.1 M BICINE pH 9.0, 2.0 M Sodium chloride
SDP
Catalog No. 501994
Encompass
HR2-932-B6/Grid Screen Sodium Chloride #B6/Grid Screen Salt HT (G12), 185 ml - 0.1 M BICINE pH 9.0, 2.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-199-4 Supplier Hampton Research Supplier No. HR2932B6
HR2-932-B6/Grid Screen Sodium Chloride #B6/Grid Screen Salt HT (G12), 185 ml - 0.1 M BICINE pH 9.0, 2.0 M Sodium chloride. Please note items from this supplier are non-returnable.