Learn More
Hampton Research HR2-932-D1/Grid Screen Sodium Chloride #D1/Grid Screen Salt HT (H07), 185 ml - 0.1 M Citric acid pH 4.0, 4.0 M Sodium chloride
SDP
Catalog No. 502001
Encompass
HR2-932-D1/Grid Screen Sodium Chloride #D1/Grid Screen Salt HT (H07), 185 ml - 0.1 M Citric acid pH 4.0, 4.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-200-1 Supplier Hampton Research Supplier No. HR2932D1
Add to Cart
Add to Cart
HR2-932-D1/Grid Screen Sodium Chloride #D1/Grid Screen Salt HT (H07), 185 ml - 0.1 M Citric acid pH 4.0, 4.0 M Sodium chloride. Please note items from this supplier are non-returnable.