Learn More
Hampton Research HR2-932-A5/Grid Screen Sodium Chloride #A5/Grid Screen Salt HT (G05), 185 ml - 0.1 M Tris pH 8.0, 1.0 M Sodium chloride
SDP
Catalog No. 501987
Encompass
HR2-932-A5/Grid Screen Sodium Chloride #A5/Grid Screen Salt HT (G05), 185 ml - 0.1 M Tris pH 8.0, 1.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-198-7 Supplier Hampton Research Supplier No. HR2932A5
HR2-932-A5/Grid Screen Sodium Chloride #A5/Grid Screen Salt HT (G05), 185 ml - 0.1 M Tris pH 8.0, 1.0 M Sodium chloride. Please note items from this supplier are non-returnable.