Learn More
Hampton Research HR2-932-B2/Grid Screen Sodium Chloride #B2/Grid Screen Salt HT (G08), 185 ml - 0.1 M Citric acid pH 5.0, 2.0 M Sodium chloride
SDP
Catalog No. 501990
Encompass
HR2-932-B2/Grid Screen Sodium Chloride #B2/Grid Screen Salt HT (G08), 185 ml - 0.1 M Citric acid pH 5.0, 2.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-199-0 Supplier Hampton Research Supplier No. HR2932B2
HR2-932-B2/Grid Screen Sodium Chloride #B2/Grid Screen Salt HT (G08), 185 ml - 0.1 M Citric acid pH 5.0, 2.0 M Sodium chloride. Please note items from this supplier are non-returnable.