Learn More
Hampton Research HR2-947-B1/Grid Screen Sodium Malonate #B1/Grid Screen Salt HT (C7), 185 ml - 1.0 M Sodium malonate pH 5.0
SDP
Supplier:  Hampton Research HR2947B1
Encompass
HR2-947-B1/Grid Screen Sodium Malonate #B1/Grid Screen Salt HT (C7), 185 ml - 1.0 M Sodium malonate pH 5.0. Please note items from this supplier are non-returnable.
Catalog No. 50-227-6