Learn More
Hampton Research HR2-947-C4/Grid Screen Sodium Malonate #C4/Grid Screen Salt HT (D4), 185 ml - 2.4 M Sodium malonate pH 6.0
SDP
Supplier:  Hampton Research HR2947C4
Encompass
HR2-947-C4/Grid Screen Sodium Malonate #C4/Grid Screen Salt HT (D4), 185 ml - 2.4 M Sodium malonate pH 6.0. Please note items from this supplier are non-returnable.
Catalog No. 50-228-5