Learn More
Electron Microscopy Sciences Gridded Microscope Slides Poly-L-Lysine
SDP
Supplier:  Electron Microscopy Sciences 6340601
Encompass_Preferred
These 3x1" slides have an indexed grid on the reverse side of the frosted end. The ink is resistant to stains and common laboratory solvents. Each square grid fills one low-powered-field (approximately 100X magnification). These slides make it easy to locate an organism or cell every time. There are eighty one squares in total. The grid measures 2 mm square-16 mm total. Numbers 1-8 and alpha A-H.
Catalog No. 50-264-09