Learn More
Electron Microscopy Sciences Gridded Microscope Slides Round Well
SDP
Catalog No. 5026413
Encompass_Preferred
These 3x1" slides have an indexed grid on the reverse side of the frosted end. The ink is resistant to stains and common laboratory solvents. Each square grid fills one low-powered-field (approximately 100X magnification). These slides make it easy to locate an organism or cell every time. There are eighty one squares in total. The grid measures 2 mm square-16 mm total. Numbers 1-8 and alpha A-H.
Catalog No. 50-264-13 Supplier Electron Microscopy Sciences Supplier No. 6340801
These 3x1" slides have an indexed grid on the reverse side of the frosted end. The ink is resistant to stains and common laboratory solvents. Each square grid fills one low-powered-field (approximately 100X magnification). These slides make it easy to locate an organism or cell every time. There are eighty one squares in total. The grid measures 2 mm square-16 mm total. Numbers 1-8 and alpha A-H.