Learn More
LabScientific Grid Slides without black background, 72/pk. 100x magnification. Black grid printed on reverse side. Ink resistant to solvents.
Catalog No. NC1807538
Encompass
Grid Slides coated with Poly-L-Lysine. You can note exactly where an organism is located on the microscopic custom grid printed on the reverse side. Each square grid fills one low powered field - 100x magnification. Ink is resistant to stains and common laboratory solvents. Black grid on the reverse side of the slide. Packaging: 72 slides/pk. Size (per gross): 25g.
Catalog No. NC1807538 Supplier LabScientific Supplier No. 4455
Grid Slides coated with Poly-L-Lysine. You can note exactly where an organism is located on the microscopic custom grid printed on the reverse side. Each square grid fills one low powered field - 100x magnification. Ink is resistant to stains and common laboratory solvents. Black grid on the reverse side of the slide. Packaging: 72 slides/pk. Size (per gross): 25g.