Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 300 µm hole, Gold
SDP
Catalog No. 501901291
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 300 um.
Material: Gold
Catalog No. 50-190-1291 Supplier Electron Microscopy Sciences Supplier No. GA300AU
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 300 um.
Material: Gold