Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 300 µm hole, Nickel
SDP
Catalog No. 501901290
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 300 um.
Material: Nickel
Catalog No. 50-190-1290 Supplier Electron Microscopy Sciences Supplier No. GA300NI
Add to Cart
Add to Cart
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 300 um.
Material: Nickel