Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 800 µm hole, Gold
SDP
Catalog No. 501901298
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 800 um.
Material: Gold
Catalog No. 50-190-1298 Supplier Electron Microscopy Sciences Supplier No. GA800AU
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 800 um.
Material: Gold