Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 200 µm hole, Nickel
SDP
Catalog No. 501901288
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 200 um.
Material: Nickel
Catalog No. 50-190-1288 Supplier Electron Microscopy Sciences Supplier No. GA200NI
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 200 um.
Material: Nickel