Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 75 µm hole, Gold
SDP
Catalog No. 501901282
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 75 um.
Material: Gold
Catalog No. 50-190-1282 Supplier Electron Microscopy Sciences Supplier No. GA75AU
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 75 um.
Material: Gold