Learn More
Electron Microscopy Sciences Grid-Stick Kit
SDP
Supplier:  Electron Microscopy Sciences 71175
Encompass_Preferred
Grid-Stick is a device for multi grid staining that is coated with a specially-formulated pressure-sensitive adhesive resistant to hold the grids in place during staining, emulsion coating, carbon coating, shadow casting, serial section collection but will not remain on the grid when removed from the stick. It is made from a thin, rigid alloy that does not react with common organic solvents or stains. Measures 4 mm wide, 75 mm long and has a slot along center with small undercut notches on one side for simple grid removal. Kit: 5 coated grid sticks, 10 staining pipettes, 2 flow-limiting plugs and bulbs.
Catalog No. 50-930-881