Learn More
Alkali Scientific ASI™ Grid Sticker, Tetron, 10mm, 200/Cs, 60mm
SDP
Catalog No. 503027675
Encompass_Preferred
ASI™ has released Grid stickers as an exclusive accessory for use with petri dish. The sticker is made of transparent label and can be attached swiftly and neatly to the bottom of petri dish.It is designed as 1 mm- and 10 mm- interval grid pattern, and can be applied to various experiments, including colony counting, plant growth measurement, and clear zone measurement for fungal test. ▪ For 90~100 mm petri dish▪ For 60 mm petri dish▪ Thickness: 70 ㎛▪ Labels remain firmly attached at temp. from 4 to 40°C
Catalog No. 50-302-7675 Supplier Alkali Scientific Supplier No. GS0060
ASI™ has released Grid stickers as an exclusive accessory for use with petri dish. The sticker is made of transparent label and can be attached swiftly and neatly to the bottom of petri dish.It is designed as 1 mm- and 10 mm- interval grid pattern, and can be applied to various experiments, including colony counting, plant growth measurement, and clear zone measurement for fungal test. ▪ For 90~100 mm petri dish▪ For 60 mm petri dish▪ Thickness: 70 ㎛▪ Labels remain firmly attached at temp. from 4 to 40°C