Learn More
Electron Microscopy Sciences TEM Grid Storage Box 100-Capacity
SDP
Supplier:  Electron Microscopy Sciences 71140
Encompass_Preferred
100 grids can be stored in identified diamond-shaped wells for daily handling or long-term storage. Complete with grid recording card.
Catalog No. 50-930-847