Learn More
Electron Microscopy Sciences Grid Storage Box 50-Capacity-Clear Cover
SDP
Catalog No. 50930857
Encompass_Preferred
Storage for 50 grids in deep diamond-shaped wells. All wells are identified. The base is resistant to organics and reactions can be carried out on grid-mounted samples in the wells. Complete with grid recording card.
Catalog No. 50-930-857 Supplier Electron Microscopy Sciences Supplier No. 71150
Storage for 50 grids in deep diamond-shaped wells. All wells are identified. The base is resistant to organics and reactions can be carried out on grid-mounted samples in the wells. Complete with grid recording card.