Learn More
Electron Microscopy Sciences EMS25 TEM Grid Storage Box, Numbered 25 Slots
SDP
Catalog No. 501929776
Encompass_Preferred
The EMS25 is a small capacity grid storage box, designed as a low cost alternative to larger grid boxes where extra storage capacity is not required. The box has a sliding cover, allowing access to 5 slots at a time. Each diamond shaped hole is capable of storing a 3.05 mm or 2.3 mm diameter grid. Storage referencing is managed via recessed alpha numeric indexing on the sides of the box.
The box contains two side features, allowing for easier handling during specimen removal, as well as a matt strip for extra notation. The boxes come with a durable 25 column card for record keeping. Our grid boxes are intended for use in both routine grid handling and for long term grid storage. Their compact flat shape and low cost make them well suited for the transportation of coated grid products and for the long term storage of catalogued specimens.
Capacity: 25 slots
Catalog No. 50-192-9776 Supplier Electron Microscopy Sciences Supplier No. 7115930
The EMS25 is a small capacity grid storage box, designed as a low cost alternative to larger grid boxes where extra storage capacity is not required. The box has a sliding cover, allowing access to 5 slots at a time. Each diamond shaped hole is capable of storing a 3.05 mm or 2.3 mm diameter grid. Storage referencing is managed via recessed alpha numeric indexing on the sides of the box.
The box contains two side features, allowing for easier handling during specimen removal, as well as a matt strip for extra notation. The boxes come with a durable 25 column card for record keeping. Our grid boxes are intended for use in both routine grid handling and for long term grid storage. Their compact flat shape and low cost make them well suited for the transportation of coated grid products and for the long term storage of catalogued specimens.
Capacity: 25 slots