Learn More
Electron Microscopy Sciences TEM Window Pure Silicon, (8) 50 µm, (1) 50x100 µm, 5 nm
SDP
Catalog No. 501929464
Encompass_Preferred
Pure Silicon TEM Windows feature imaging windows with 5 to 15 nm thickness, reducing background contribution and interference for higher contrast imaging. The membrane can be vigorously plasma cleaned to remove organic contamination, unlike traditional carbon grids. The elemental silicon composition of TEM Windows remarkably increases stability at high beam currents and at high annealing temperatures. With a minimal background signal, elemental analyses of samples containing nitrogen and/or carbon is possible by EDX and EELS. Nanoporous with pores from 10-60nm.
Dimensions: (8) 50 µm, (1) 50x100 µm
Thickness: 5 nm
Catalog No. 50-192-9464 Supplier Electron Microscopy Sciences Supplier No. 7604272
Pure Silicon TEM Windows feature imaging windows with 5 to 15 nm thickness, reducing background contribution and interference for higher contrast imaging. The membrane can be vigorously plasma cleaned to remove organic contamination, unlike traditional carbon grids. The elemental silicon composition of TEM Windows remarkably increases stability at high beam currents and at high annealing temperatures. With a minimal background signal, elemental analyses of samples containing nitrogen and/or carbon is possible by EDX and EELS. Nanoporous with pores from 10-60nm.
Dimensions: (8) 50 µm, (1) 50x100 µm
Thickness: 5 nm