Learn More
Electron Microscopy Sciences Grid Transporting Box
SDP
Catalog No. 501901302
Encompass_Preferred
This unique Box allows for the storage and transportation of grids without any worry of them moving around or being damaged. The Plate is made from Silicone and has 54 individual compartments 10 mm in size. Each compartment can hold 3 grids and the grids can be easily picked up from the Silicone surface.
Catalog No. 50-190-1302 Supplier Electron Microscopy Sciences Supplier No. 7117301
This unique Box allows for the storage and transportation of grids without any worry of them moving around or being damaged. The Plate is made from Silicone and has 54 individual compartments 10 mm in size. Each compartment can hold 3 grids and the grids can be easily picked up from the Silicone surface.