Learn More
Electron Microscopy Sciences Optical Resolution Grid with line pitch 0.25 mm
SDP
Catalog No. 501901584
Encompass_Preferred
Consists of elements of 5 bars ranging from 1 line per mm to 18 lines per mm, with adjacent numbers indicating the number of lines per mm. Produced as a vacuum deposited chrome image on 75 mm x 75 mm glass.
Catalog No. 50-190-1584 Supplier Electron Microscopy Sciences Supplier No. 6809823
Add to Cart
Add to Cart
Consists of elements of 5 bars ranging from 1 line per mm to 18 lines per mm, with adjacent numbers indicating the number of lines per mm. Produced as a vacuum deposited chrome image on 75 mm x 75 mm glass.