Learn More
Electron Microscopy Sciences Optical Resolution Grid with line pitch of 0.5 mm
SDP
Catalog No. 501901583
Encompass_Preferred
Consists of elements of 5 bars ranging from 1 line per mm to 18 lines per mm, with adjacent numbers indicating the number of lines per mm. Produced as a vacuum deposited chrome image on 75 mm x 75 mm glass.
Catalog No. 50-190-1583 Supplier Electron Microscopy Sciences Supplier No. 6809822
Consists of elements of 5 bars ranging from 1 line per mm to 18 lines per mm, with adjacent numbers indicating the number of lines per mm. Produced as a vacuum deposited chrome image on 75 mm x 75 mm glass.