Learn More
Electron Microscopy Sciences Gold Wire, 0.20 mm, Purity 99.99% 10 FT
SDP
Catalog No. NC1433031
Encompass_Preferred
High purity, vacuum grade. These metal wires are suitable for scanning and electron microscopy techniques.
Size: 0.20 mm
Purity: 99.99%
Catalog No. NC1433031 Supplier Electron Microscopy Sciences Supplier No. 73100
High purity, vacuum grade. These metal wires are suitable for scanning and electron microscopy techniques.
Size: 0.20 mm
Purity: 99.99%