Learn More
Ibidi USA Gridded Glass Coverslips Grid-50, 100/box
SDP
Catalog No. NC1872351
Encompass
Grid repeat distance: 50 µm. Diameter: 28 mm. High-quality glass coverslip
This item is not returnable. View return policy
Catalog No. NC1872351 Supplier Ibidi USA Supplier No. 10817
Grid repeat distance: 50 µm. Diameter: 28 mm. High-quality glass coverslip