Learn More
Grainger Griddle Clnr Degreaser Packet 3.2oz PK40
Catalog No. 502767841
Encompass_Preferred
The Grainger 16Z997 is a Westward brand 1/4-inch drive socket set. It contains 14 SAE sockets ranging from 1/4 inch to 9/16 inch and 14 metric sockets ranging from 4mm to 14mm. The chrome vanadium sockets have a knurled design for oil and slip resistance. This 108-piece set comes organized in a molded case for convenient storage and portability. The sockets have a matte finish and meet or exceed ANSI standards. This is an ideal starter socket set for automotive and machinery repairs.
Catalog No. 50-276-7841 Supplier Grainger Supplier No. 16Z997
Add to Cart
Add to Cart
The Grainger 16Z997 is a Westward brand 1/4-inch drive socket set. It contains 14 SAE sockets ranging from 1/4 inch to 9/16 inch and 14 metric sockets ranging from 4mm to 14mm. The chrome vanadium sockets have a knurled design for oil and slip resistance. This 108-piece set comes organized in a molded case for convenient storage and portability. The sockets have a matte finish and meet or exceed ANSI standards. This is an ideal starter socket set for automotive and machinery repairs.