Learn More
Electron Microscopy Sciences Athene Hexagonal, 100 mesh Grid, Nickel
SDP
Catalog No. 501926668
Encompass_Preferred
Athene grids are renowned for the highest quality standards, exceptionally refined grid bars, and good handling characteristics. 3.05 mm diameter.
Grid: Nickel
Catalog No. 50-192-6668 Supplier Electron Microscopy Sciences Supplier No. AH100NI
Athene grids are renowned for the highest quality standards, exceptionally refined grid bars, and good handling characteristics. 3.05 mm diameter.
Grid: Nickel