Learn More
Electron Microscopy Sciences Gilder High Mesh Grids Circular 2000 mesh Copper Grid
SDP
Catalog No. 501926643
Encompass_Preferred
A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18um) thickness. Mesh diameter: 2 mm.
Grid: Copper
Pitch: 12.5 µm
Holes: 6.5 µm
Catalog No. 50-192-6643 Supplier Electron Microscopy Sciences Supplier No. G2000HACU
A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18um) thickness. Mesh diameter: 2 mm.
Grid: Copper
Pitch: 12.5 µm
Holes: 6.5 µm