Learn More
Cayman Chemical GrIes Reagent R2 10ea
SDP
Catalog No. 502896656
Encompass_Preferred
Griess Reagent component for determination of nitrite in aqueous solutions.
Catalog No. 50-289-6656 Supplier Cayman Chemical Supplier No. 78002010 EA
Griess Reagent component for determination of nitrite in aqueous solutions.