Learn More
Enzo Life Sciences Griess Reagent (50ml)
SDP
Catalog No. 502009067
Encompass_Preferred
Reagent for the determination of nitrite (NO2-) in biological media. Consists of 1 part 0.1% naphthylethylenediamine dihydrochloride in distilled water plus 1 part 1% sulfanilamide (or sulfanilic acid) in 25-50% acetic acid. Appearance: Colorless to slightly pink liquid. Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9067 Supplier Enzo Life Sciences Supplier No. ALX400004L050
Add to Cart
Add to Cart
Reagent for the determination of nitrite (NO2-) in biological media. Consists of 1 part 0.1% naphthylethylenediamine dihydrochloride in distilled water plus 1 part 1% sulfanilamide (or sulfanilic acid) in 25-50% acetic acid. Appearance: Colorless to slightly pink liquid. Solubility: Soluble in water. Long Term Storage: +4°C.