Learn More
Sigma Aldrich Fine Chemicals Biosciences Griess reagent modified10G
Catalog No. 502900868
Encompass
Griess reagent is based on Griess reaction. The components include naphthylethylenediamine dihydrochloride suspended in water and sulphanilamide in phosphoric acid. This reagent reacts with nitrite in samples to form a purple azo product absorbance of which is measured at 546 nm.
Catalog No. 50-290-0868 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G441010G
Griess reagent is based on Griess reaction. The components include naphthylethylenediamine dihydrochloride suspended in water and sulphanilamide in phosphoric acid. This reagent reacts with nitrite in samples to form a purple azo product absorbance of which is measured at 546 nm.