Learn More
Chemglass Life Sciences Beaker, Heavy Duty, 400mL, Griffin, Double Scale, Graduated
SDP
Catalog No. NC1319361
Encompass_Preferred
Beaker, Heavy Duty, 400mL, Griffin, Double Scale, Graduated. Heavy duty beakers specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. For convenience, the 250 through 4L beakers have a double graduated metric scale to indicate their approximate content. All sizes have an extra large marking spot. Reference: ASTM E-960.
Catalog No. NC1319361 Supplier Chemglass Life Sciences Supplier No. CG8050400
Beaker, Heavy Duty, 400mL, Griffin, Double Scale, Graduated. Heavy duty beakers specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. For convenience, the 250 through 4L beakers have a double graduated metric scale to indicate their approximate content. All sizes have an extra large marking spot. Reference: ASTM E-960.