Learn More
Humboldt Griffin-Type Glass Tubing Cutter
SDP
Catalog No. 11340
Change view
Click to view available options
Griffin-type Glass Tubing Cutter; Cuts glass tubing up to 40mm
Catalog No.
11-340 Griffin-type Glass Tubing Cutter; Cuts glass tubing up to 40mm
1 options
This item is not returnable. View return policy
Catalog No. 11-340 Supplier Humboldt Supplier No. H13520
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Strong cutter is corrosion resistant
  • Cuts glass tubing up to 40mm
  • Nickel-plated, zinc alloy, die cast
Specifications
Material Zinc Alloy
For Use With (Application) Cuts glass tubing up to 40mm
Product Type Glass Tubing Cutter