Learn More
Bioworld Griffonia (Bandeiraea) simplicifolia Lectin (GSL I+II) - Rhodamine (TRITC)
Catalog No. 50199137
Encompass
Technical Specifications:Ligand Source:Griffonia simplicifoliaMolecular Weight:114 kDaCarbohydrate Specificity:αGal, αGalNAcInhibiting/Eluting Sugar: Galactose/N-AcetylgalactoseCarbohydrate for Elution:0.2M Galactose/N-AcetylgalactoseBlood Group Specificity:B>>A1Conjugate: RhodamineForm:LiquidIsoelectric (pI):5-6.5Storage: -20°CApplication note: Calcium and Manganese are required for binding.
Catalog No. 50-199-137 Supplier Bioworld Supplier No. 217610281
Technical Specifications:Ligand Source:Griffonia simplicifoliaMolecular Weight:114 kDaCarbohydrate Specificity:αGal, αGalNAcInhibiting/Eluting Sugar: Galactose/N-AcetylgalactoseCarbohydrate for Elution:0.2M Galactose/N-AcetylgalactoseBlood Group Specificity:B>>A1Conjugate: RhodamineForm:LiquidIsoelectric (pI):5-6.5Storage: -20°CApplication note: Calcium and Manganese are required for binding.