Learn More
Strem, An Ascensus Company CAS 920-39-8. 1mole. i-Propylmagnesium bromide 2.9M (35wt% 1wt%) in 2-methyltetrahydrofuran. MFCD00672110
SDP
Catalog No. 502110706
Encompass
CAS 920-39-8. 1mole. i-Propylmagnesium bromide 2.9M (35wt% 1wt%) in 2-methyltetrahydrofuran. MFCD00672110. Molecular Weight 147.30. Molecular Formula (CH3)2CHMgBr. Color/form liq. Strem 12-0825. http//www.strem.com/catalog/v/12-0825/
Catalog No. 50-211-0706 Supplier Strem, An Ascensus Company Supplier No. 1208251MOL
CAS 920-39-8. 1mole. i-Propylmagnesium bromide 2.9M (35wt% 1wt%) in 2-methyltetrahydrofuran. MFCD00672110. Molecular Weight 147.30. Molecular Formula (CH3)2CHMgBr. Color/form liq. Strem 12-0825. http//www.strem.com/catalog/v/12-0825/