Learn More
Aviva Systems Biology Corp GRIK4 Antibody - N-terminal region (AVARP13040_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13040_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GRIK4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GRIK4. Alternate names: KA1, EAA1, GRIK, GluK4ApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53783