Learn More
Aviva Systems Biology Corp GRIK5 Antibody (ARP37674_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37674_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRIK5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GRIK5. Alternate names: EAA2, GRIK2, KA2, GluK5ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56390