Learn More
Aviva Systems Biology Corp Grik5 Antibody - N-terminal region (ARP35064_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35064_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Grik5. It was validated on Western Blot. . Alternate names: GluRgamma2, KA2, MGC118086ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-441