Learn More
Aviva Systems Biology Corp GRIN2A Antibody - middle region (ARP35012_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35012_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRIN2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GRIN2A. Alternate names: NMDAR2A, NR2A, EPND, GluN2AApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55629