Learn More
Fritsch Milling And Sizing Inc Grinding Ball 20 mm Hardmetal Tungsten Carbide
SDP
Catalog No. 501534688
Encompass
Grinding ball 20 mm hardmetal tungsten carbide
General material specification:
Material: hardmetal tungsten carbide - WC
Abrasion resistance: very good
Suitable for: hard, abrasive samples
Catalog No. 50-153-4688 Supplier Fritsch Milling And Sizing Inc Supplier No. 55.0200.08
Grinding ball 20 mm hardmetal tungsten carbide
General material specification:
Material: hardmetal tungsten carbide - WC
Abrasion resistance: very good
Suitable for: hard, abrasive samples