Learn More
Fritsch Milling And Sizing Inc Grinding Ball 20 mm Hardened, Stainless Steel
SDP
Catalog No. 501534689
Encompass
Grinding ball 20 mm hardened, stainless steel
General material specification:
Material: hardened, stainless steel - Fe-Cr
Abrasion resistance: good
Suitable for: hard, medium-hard, brittle samples
Catalog No. 50-153-4689 Supplier Fritsch Milling And Sizing Inc Supplier No. 55.0200.09
Grinding ball 20 mm hardened, stainless steel
General material specification:
Material: hardened, stainless steel - Fe-Cr
Abrasion resistance: good
Suitable for: hard, medium-hard, brittle samples