Learn More
Fritsch Milling And Sizing Inc Grinding Ball 50 mm Hardened Steel, P-0
SDP
Catalog No. 501534359
Encompass
General material specification:
Material: hardened steel
Abrasion resistance: good
Suitable for: medium-hard, brittle samples
Catalog No. 50-153-4359 Supplier Fritsch Milling And Sizing Inc Supplier No. 40.0190.09
General material specification:
Material: hardened steel
Abrasion resistance: good
Suitable for: medium-hard, brittle samples