Learn More
Fritsch Milling And Sizing Inc Grinding Ball 50 mm Hardmetal Tungsten Carbide, P-0
SDP
Catalog No. 501534360
Encompass
General material specification:
Material: hardmetal tungsten carbide - WC - GC10
Abrasion resistance: very good
Suitable for: hard, abrasive samples
Catalog No. 50-153-4360 Supplier Fritsch Milling And Sizing Inc Supplier No. 40.0200.08
General material specification:
Material: hardmetal tungsten carbide - WC - GC10
Abrasion resistance: very good
Suitable for: hard, abrasive samples