Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ops Diagnostics GRINDING BALLS 3 MM 440C SS

Catalog No. NC2021788
Encompass

Grinding Balls, 3 mm, 440C Stainless Steel, Bottle of 2500

Catalog No. NC2021788 Supplier Ops Diagnostics Supplier No. GBSS 118250009
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Grinding Balls, 3 mm, 440C Stainless Steel, Bottle of 2500

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.