Learn More
Fritsch Milling And Sizing Inc Grinding Bowl 250 Ml Easy-Gtm Hardened,Stainless Steel P-7 Pl
SDP
Catalog No. 501534630
Encompass
Grinding bowl 250 ml EASY-GTM hardened,stainless steel P-7 pl for the use of the EASY GTM-System - for controlling the grinding process and continuous measurement of gas pressure and temperature.
Turn your Planetary Mill PULVERISETTE 5 premium line  into an analytical measuring system by using the EASY GTM system (Gas Pressure and Temperature Measurement System) with special lid and transmitter as well as a receiver board and the included MillControl software. Your advantage: easy and safe monitoring and analysis of thermal effects, physical and chemical reactions or increases/decreases in pressure through continuous measurement of gas pressure and temperature directly in the grinding bowl. The mill is automatically controlled to ensure that the set parameters are not exceeded.
EASY GTM grinding bowls are also available in zirconium oxide and and hardmetal tungsten carbide.
Only one receiver unit with software MillControl has to be ordered.
Catalog No. 50-153-4630 Supplier Fritsch Milling And Sizing Inc Supplier No. 50.9280.00
Add to Cart
Add to Cart
Request a Quote
Grinding bowl 250 ml EASY-GTM hardened,stainless steel P-7 pl for the use of the EASY GTM-System - for controlling the grinding process and continuous measurement of gas pressure and temperature.
Turn your Planetary Mill PULVERISETTE 5 premium line  into an analytical measuring system by using the EASY GTM system (Gas Pressure and Temperature Measurement System) with special lid and transmitter as well as a receiver board and the included MillControl software. Your advantage: easy and safe monitoring and analysis of thermal effects, physical and chemical reactions or increases/decreases in pressure through continuous measurement of gas pressure and temperature directly in the grinding bowl. The mill is automatically controlled to ensure that the set parameters are not exceeded.
EASY GTM grinding bowls are also available in zirconium oxide and and hardmetal tungsten carbide.
Only one receiver unit with software MillControl has to be ordered.